Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant IL20 (fromE. coli)
Catalog # PRSI91-129
Nome del fornitore:  ProSci Inc.
Numero CAS:  
Human recombinant IL20 (fromE. coli)
Catalog # PRSI91-129
Nome del fornitore:  ProSci Inc.
Numero CAS:  

Specifiche

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Cytokine Zcyto10|IL-20|ZCYTO10|Interleukin-20|IL20
  • Protein/peptide name:
    IL20
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    20.07 kD
  • Sequence:
    Leu25-Glu176
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifiche

Informazioni su questo articolo

Interleukin-20 (IL-20) is a member of the IL-10 family of regulatory cytokines that includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Members of this family share partial homology in their amino acid sequences but they are dissimilar in their biological functions. IL-20 exhibits approximately 28% amino acid identity with IL-10 and 76% amino acid identity with mouse IL-20. There are two heterodimeric receptor complexes for IL-20. The first is composed of IL-20 R alpha and IL-20 R beta . The second is composed of IL-22 R and IL-20 R beta . Whereas the IL-22 R/IL-20 R beta complex is shared with IL-24, the IL-20 R alpha/IL-20 R beta complex is shared with both IL-19 and IL-24. IL-20 has been shown to initiate transduction cascades involving STAT3 and stimulates the induction of pro-inflammatory genes including TNF- alpha and MCP-1. Initial functional studies using transgenic mice suggest that IL-20 has the ability to regulate skin development. The over-expression of both human and mouse forms of IL-20 results in keratinocyte hyper-proliferation, abnormal epidermal differentiation, and neonatal lethality. In humans, IL-20 and its receptors are up-regulated in psoriatic skin, and polymorphisms in the IL-20 gene have been associated with plaque-type psoriasis. IL-20 may also have a role in hematopoiesis. It enhances the proliferation of multi-potential progenitors in vitro and increases their numbers and cell cycling status in IL-20 transgenic mice. IL-20 is also shown to suppress COX-2 and PGE2 and acts as an inhibitor of angiogenesis in model systems.

  • This recombinant protein can be used for biological assays. For research use only.