Order Entry
Italy
ContactUsLinkComponent
 
Human Recombinant DHS (from E. coli)
  PRSI91-112
 :  
Human Recombinant DHS (from E. coli)
  PRSI91-112
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    DS|Deoxyhypusine Synthase|DHPS|DHS
  • Protein/peptide name:
    DHS
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    42.04 kD
  • Sequence:
    Met1-Asp369
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 20% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Human Deoxyhypusine Synthase (DHS) is vital for the first step of hypusine biosynthesis. DHS catalyzes the NAD-dependent oxidative cleavage of spermidine, the subsequent transfer of the butylamine moiety of spermidine to the epsilon-amino group of a specific lysine residue of the eIF-5A precursor protein to form the intermediate deoxyhypusine residue.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag