Order Entry
Italy
ContactUsLinkComponent
Human recombinant CRYAB (from E. coli)
Human recombinant CRYAB (from E. coli)
Catalog # PRSI91-101
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CRYAB (from E. coli)
Catalog # PRSI91-101
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Renal Carcinoma Antigen NY-REN-27|Alpha-Crystallin B Chain|CRYA2|Alpha(B)-Crystallin|Heat Shock Protein Beta-5|Rosenthal Fiber Component|HspB5|CRYAB
  • Protein/peptide name:
    CRYAB
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21.22 kD
  • Sequence:
    Met1-Lys175
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays

Specifications

About this item

alpha Crystallin B Chain (CRYAB) is a cytoplasmic protein that belongs to the small heat shock protein (HSP20) family. Alpha crystallins are composed of two gene products: alpha-A and alpha-B, for acidic and basic, respectively. Alpha crystallins can be induced by heat shock and are members of the small heat shock protein (sHSP also known as the HSP20) family. Alpha crystallins acts as molecular chaperones and hold them in in large soluble aggregates. CRYAB is expressed widely in many tissues and organs. It may contribute to the transparency and refractive index of the lens. The deficiency of CRYAB is the cause of myopathy myofibrillar type 2 (MFM2) and cataract posterior polar type 2 (CTPP2).

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag