Order Entry
Italy
ContactUsLinkComponent
Human Recombinant CCL16 (from E. coli)
Human Recombinant CCL16 (from E. coli)
  PRSI91-051
 :  
Human Recombinant CCL16 (from E. coli)
  PRSI91-051
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    NCC-4|CCL16|Chemokine LEC|ILINCK|IL-10-Inducible Chemokine|Liver-Expressed Chemokine|C-C Motif Chemokine 16|Monotactin-1|Lymphocyte and Monocyte Chemoattractant|MTN-1|SCYA16|Small-Inducible Cytokine A16|NCC4|HCC-4|Chemokine CC-4|LCC-1|LMC
  • Protein/peptide name:
    CCL16
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    11 kD
  • Sequence:
    Gln24-Gln120
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

CCL16 is a member of CC chemokine family. CCL16 cDNA encodes a 120 amino acid peptide along with a 23 amino acids signal peptide that is cleaved to generate 97 amino acid protein. CCL16 is distantly related to other CC chemokines, showing less than 30% sequence identity. CCL16 elicits its effects on cells by interacting with cell surface chemokine receptors such as CCR1, CCR2, CCR5 and CCR8. Recombinant CCL16 has been shown to chemoattract human monocytes and THP1 cells but not resting lymphocytes nor neutrophils. CCL16 has potent myelosuppressive activity, suppresses proliferation of myeloid progenitor cells. CCL16ninduces a calcium flux in THP1 cells that can be desensitized by prior exposure to RANTES, suggesting that CCL16 and RANTES share the same receptor in THP1 cells.

  • This recombinant protein can be used for biological assays. For research use only.