Order Entry
Italy
ContactUsLinkComponent
Human Recombinant CCL8 (from E. coli)
Human Recombinant CCL8 (from E. coli)
Catalog # PRSI91-050
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant CCL8 (from E. coli)
Catalog # PRSI91-050
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    HC14|SCYA8|Monocyte Chemoattractant Protein 2|MCP2|MCP-2|Monocyte Chemotactic Protein 2|SCYA10|CCL8|Small-Inducible Cytokine A8|C-C Motif Chemokine 8
  • Protein/peptide name:
    CCL8
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    8.9 kD
  • Sequence:
    Gln24-Pro99
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Human Chemokine (C-C Motif) Ligand 8 (CCL8) is produced by human MG63 osteosarcoma cells. CCL8 shares 62% and 58% amino acid sequence identity with MCP-1 and MCP-3, respectively. All three MCP proteins are monocyte chemoattractants. CCL8 is chemotactic for and activates many different immune cells, including mast cells, eosinophils and basophils, which are implicated in allergic response, and monocytes, T cells, and NK cells that are involved in the inflammatory response. CCL8 elicits its effects by binding to several different cell surface receptors including CCR1, CCR2B and CCR5.

  • This recombinant protein can be used for biological assays. For research use only.