Order Entry
Italy
ContactUsLinkComponent
Human recombinant IGF-I (4-70) (from E. coli)
Human recombinant IGF-I (4-70) (from E. coli)
Catalog # PRSI91-023
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IGF-I (4-70) (from E. coli)
Catalog # PRSI91-023
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biological activity:
    ED50 is greater than 200 ng/ml as determined by an IGF binding protein assay.
  • Protein synonyms:
    IBP1|IGF1|Mechano Growth Factor|IGF-I|Insulin-like Growth Factor I|MGF|Somatomedin-C
  • Protein/peptide name:
    IGF-I (4-70)
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    7.6 kD
  • Sequence:
    Gly49-Ala118
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 300mM NaAc, pH 6.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in 500mM Acetic Acid. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Insulin-like growth factor I (IGF1) belongs to the family of insulin-like growth factors that are structurally homologous to proinsulin. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contains the N- and C-terminal propeptide regions. Mature human IGF-I consisting of 70 amino acids has 94% identity with mouse IGF-I and exhibits cross-species activity. IGF-1 binds IGF-IR, IGF-IIR, and the insulin receptor and plays a key role in cell cycle progression, cell proliferation and tumor progression. IGF-1 expression is regulated by growth hormone. R3 IGF-1 is an 83 amino acid analog of IGF-1 comprising the complete human IGF-1 sequence with the substitution of an Arg (R) for the Glu(E) at position three, hence R3, and a 13 amino acid extension peptide at the N terminus. R3 IGF-1 has been produced with the purpose of increasing biological activity. R3 IGF-1 is significantly more potent than human IGF-I in vitro.

  • This recombinant protein can be used for biological assays. For research use only.