Order Entry
Italy
ContactUsLinkComponent
Human recombinant neprilysin (from HEK293 cells)
Human recombinant neprilysin (from HEK293 cells)
  PRSI96-540
 :  
undefined
Human recombinant neprilysin (from HEK293 cells)
  PRSI96-540
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein synonyms:
    NEP|CD10|DKFZp686O16152|CALLA|MGC126681|MME|MGC126707|SFE|Neprilysin
  • Protein/peptide name:
    Neprilysin
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    80 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 150 mM NaCl, pH8.0
  • Tested applications:
    Western Blot

 

 

Cluster of differentiation 10 (CD10) is also known as membrane metallo-endopeptidase, neutral endopeptidase (NEP), Neprilysin, and common acute lymphoblastic leukemia antigen (CALLA), is a 90-110-kDa type II transmembrane glycoprotein normally expressed by a variety of tissues, including epithelial cells of the prostate, kidney, intestine, endometrium, adrenal glands, and lung. This zinc-dependent metalloprotease enzyme cleaves peptide bonds on the amino side of hydrophobic residues and inactivates a variety of physiologically active secreted peptides. CD20 is thought to be the rate-limiting degrading enzyme of amyloid β peptide (Aβ) whose abnormal misfolding and aggregation in neural tissue has been implicated in the development of Alzheimer's disease (AD). CD10 is also identified as the common acute lymphoblastic leukemia antigen (CALLA) present on leukemic cells of pre-B phenotype, and thus serves as the most important biomarker in the diagnosis of human acute lymphocytic leukemia (ALL).