Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant interleukin 29 (from HEK293 cells)
Catalog # PRSI96-451
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant interleukin 29 (from HEK293 cells)
Catalog # PRSI96-451
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured in an antiviral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. The ED50 for this effect is typically 15 ng/mL.
  • Protein synonyms:
    IFNL1|IL29|IFNλ1|IFN lambda 1|Interleukin-29
  • Protein/peptide name:
    Interleukin 29
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    21.1 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item