Recombinant Glyco A33inan (from HEK293 cells)
Catalog # PRSI96-364
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Protein synonyms:GPA33|A33
- Protein/peptide name:Glyco A33inan
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:24.7 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
Specifications
About this item
Glycoprotein A33 (GPA33) is also known as Cell surface A33 antigen, is a single-pass type I membrane protein which is expressed in normal gastrointestinal epithelium and in 95% of colon cancers. GPA33 The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 1 Ig-like C2-type (immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily, which contains. GPA33 may play a role in cell-cell recognition and signaling.