Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant Igsf161 (from HEK293 cells)
Catalog # PRSI96-158
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant Igsf161 (from HEK293 cells)
Catalog # PRSI96-158
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein synonyms:
    CLM-6|CMRF35-A1|CMRF-35|CD300C|CMRF35A|IGSF16
  • Protein/peptide name:
    Igsf161
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    18.7 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

CMRF35-like molecule 6 (CLM-6) is also known as CD300 antigen-like family member C, CMRF35-A1, Immunoglobulin superfamily member 16 (IgSF16) , CD antigen CD300c, which belongs to the CD300 family. CLM-6 contains one Ig-like V-type (immunoglobulin-like) domain. CD300a/c RNA and surface expression were downregulated after stimulation of plasmacytoid dendritic cells (pDCs) with TLR7 and TLR9 ligands. Exogenous interferon (IFN)-alpha down-regulated CD300a/c expression. CD300a and CD300c play an important role in the cross-regulation of TNF-alpha and IFN-alpha secretion from pDCs. CD300a and CD300c are indistinguishable on the surface of NK cells.