Human recombinant Adam8 (from HEK293 cells)
Catalog # PRSI96-013
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:CD156a|MS2|ADAM8
- Protein/peptide name:ADAM8
- Purity:>85% as determined by SDS-PAGE.
- Molecular weight:71.6 kDa
- Formulation:Lyophilized, 50 mM Tris, 150 mM NaCl, 10 mM CaCl2, pH7.5
- Tested applications:Western Blot
Specifications
About this item
Disintegrin and metalloproteinase domain-containing protein 8 is also known as ADAM8, Cell surface antigen MS2 and CD antigen CD156a, which belongs to the ADAM (a disintegrin and metalloprotease domain) family. ADAM family play a fundamental role in diverse processes such as asthma, development, angiogenesis and cancer through their activities in cell adhesion/fusion, membrane protein shedding, and signal transduction. ADAM8 / CD156a contains 1 disintegrin domain, 1 EGF-like domain and 1 peptidase M12B domain. ADAM8 / MS2 / CD156a is expressed on neutrophils and monocytes. ADAM8 / MS2 possible involvement in extravasation of leukocytes.