Order Entry
Italy
ContactUsLinkComponent
Human caspase 3 peptide (17 aa in the center)
Human caspase 3 peptide (17 aa in the center)
Catalog # PRSI3445P
Supplier:  ProSci Inc.
CAS Number:  
Human caspase 3 peptide (17 aa in the center)
Catalog # PRSI3445P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    17 aa in the center
  • Protein/peptide name:
    Caspase 3 Peptide (17 aa in the center)
  • Tested applications:
    Blocking

Specifications

About this item

Caspase-3 peptide is used for blocking the activity of Caspase-3 antibody.