Order Entry
Italy
ContactUsLinkComponent
 
Human PRICKLE1 peptide (19 aa near C-terminus)
  PRSI6561P
 :  
Human PRICKLE1 peptide (19 aa near C-terminus)
  PRSI6561P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    19 aa near C-terminus
  • Protein/peptide name:
    PRICKLE1 Peptide (19 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

PRICKLE1 peptide is used for blocking the activity of PRICKLE1 antibody.