Order Entry
Italy
ContactUsLinkComponent
 
Human of GOLPH2 peptide (18 aa in the center)
  PRSI5441P
 :  
Human of GOLPH2 peptide (18 aa in the center)
  PRSI5441P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    18 aa in the center
  • Protein/peptide name:
    of GOLPH2 Peptide (18 aa in the center)
  • Tested applications:
    Blocking

 

 

GOLPH2 peptide is used for blocking the activity of GOLPH2 antibody.