Order Entry
Italy
ContactUsLinkComponent
OCIAD2 Peptide (17 aa near N-terminus)
OCIAD2 Peptide (17 aa near N-terminus)
Catalog # PRSI5465P
Supplier:  ProSci Inc.
CAS Number:  
OCIAD2 Peptide (17 aa near N-terminus)
Catalog # PRSI5465P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    17 aa near N-terminus
  • Protein/peptide name:
    OCIAD2 Peptide (17 aa near N-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

OCIAD2 peptide is used for blocking the activity of OCIAD2 antibody.