Order Entry
Italy
ContactUsLinkComponent
 
OCIAD1 Peptide (16 aa near N-terminus)
Catalog # PRSI5461P
Supplier:  ProSci Inc.
CAS Number:  
OCIAD1 Peptide (16 aa near N-terminus)
Catalog # PRSI5461P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    16 aa near N-terminus
  • Protein/peptide name:
    OCIAD1 Peptide (16 aa near N-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

OCIAD1 peptide is used for blocking the activity of OCIAD1 antibody.