Order Entry
Italy
ContactUsLinkComponent
FRMPD1 peptide (15 aa near C-terminus)
FRMPD1 peptide (15 aa near C-terminus)
Catalog # PRSI5453P
Supplier:  ProSci Inc.
CAS Number:  
FRMPD1 peptide (15 aa near C-terminus)
Catalog # PRSI5453P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near C-terminus
  • Protein/peptide name:
    FRMPD1 Peptide (15 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

FRMPD1 peptide is used for blocking the activity of FRMPD1 antibody.