Human Gle1 peptide (15 aa in the center)
Catalog # PRSI4973P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa in the center
- Protein/peptide name:Gle1 Peptide (15 aa in the center)
- Tested applications:Blocking
Specifications
About this item
Gle1 peptide is used for blocking the activity of Gle1 antibody.