Human ASAH2 peptide (17 aa near C-terminus)
Catalog # PRSI4743P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa near C-terminus
- Protein/peptide name:ASAH2 Peptide (17 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
ASAH2 peptide is used for blocking the activity of ASAH2 antibody.