Order Entry
Italy
ContactUsLinkComponent
 
Mouse TSLP receptor peptide (21 aa in the center)
  PRSI4209P
 :  
undefined
Mouse TSLP receptor peptide (21 aa in the center)
  PRSI4209P
 :  

 

  • Pk:
    50 µG
  • Species:
    Mouse
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    21 aa in the center
  • Protein/peptide name:
    TSLP receptor Peptide (21 aa in the center)
  • Tested applications:
    Blocking

 

 

TSLP receptor peptide is used for blocking the activity of TSLP receptor antibody.