Order Entry
Italy
ContactUsLinkComponent
Human TLR1 peptide (17 aa in the center)
Human TLR1 peptide (17 aa in the center)
Catalog # PRSI3641P
Supplier:  ProSci Inc.
CAS Number:  
Human TLR1 peptide (17 aa in the center)
Catalog # PRSI3641P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    17 aa in the center
  • Protein/peptide name:
    TLR1 Peptide (17 aa in the center)
  • Tested applications:
    Blocking

Specifications

About this item

TLR1 peptide is used for blocking the activity of TLR1 antibody.