Human Bak peptide (14 aa near N-terminus)
PRSI3347P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa near N-terminus
- Protein/peptide name:Bak Peptide (14 aa near N-terminus)
- Tested applications:Blocking
Bak peptide is used for blocking the activity of Bak antibody.