Mouse TLR7 peptide (14 aa in the center)
Catalog # PRSI3269P
Nome del fornitore: ProSci Inc.
Numero CAS:
Specifiche
- Pk:50 µG
- Species:Mouse
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa in the center
- Protein/peptide name:TLR7 Peptide (14 aa in the center)
- Tested applications:Blocking
Specifiche
Informazioni su questo articolo
TLR7 peptide is used for blocking the activity of the TLR7 antibody.