Anti-CXCL2 Rabbit Polyclonal Antibody (Biotin)
PRSIXP-5237BT
- Antigen name:Chemokine (C-X-C motif) Ligand 2
- Antigen symbol:CXCL2
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:MIP-2|CINC-2a|GROb|MGSA-b|MIP-2a|GRO2|SCYB2|CINC-3|MIP2|GRO beta|MIP2A
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mMIP-2 (murine MIP-2).
- Size:50 µg
- Pk:50 µG
Anti-mMIP-2 specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CXCL2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: