Anti-CCL4 Rabbit Polyclonal Antibody
PRSIXP-5235
- Antigen name:Chemokine (C-C motif) ligand 4
- Antigen symbol:CCL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonyms:SCYA4|HC21|G-26|AT744.1|LAG-1|MIP-1-beta|SCYA2|ACT2|MIP1B|MIP1B1|LAG1
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hMIP-1-beta.
- Size:100 µg
- Pk:100 µG
Anti-hMIP-1-beta specific antibody was purified by affinity chromatography employing immobilized hMIP-1-beta matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CCL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human