Specifications
- Antigen name:Chemokine (C-X-C motif) Ligand 10
- Antigen symbol:CXCL10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonyms:IFI10|SCYB10|C7|crg-2|INP10|gIP-10|IP-10|mob-1
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIP-10 (human IP-10).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Interferon-gamma-inducible 10 kD protein (IP-10), is a CXC chemokine with chemoattractant properties for CD4-positive T cells and inhibits early normal and leukemic hemopoietic progenitor proliferation. IP-10 is produced by a wide variety of cell types ranging from neutrophils and monocytes to hepatocytes, endothelial cells and keratinocytes. The cytokine is reported to be involved in a scala of inflammatory pathologies such as HIV encephalitis, cutaneous T cell lymphoma, chronic hepatitis and acute anterior uveitis. Various observations strongly suggest a role for the CXC chemokines IL-8 and IP-10 in the regulation of angiogenic activity in cancer and in idiopathic pulmonary fibrosis.
Anti-hIP-10 specific antibody was purified by affinity chromatography employing immobilized hIP-10 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CXCL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: