Anti-GPR98 Rabbit Polyclonal Antibody
Catalog # PRSI71-210
Supplier: ProSci Inc.
Specifications
- Antigen name:G Protein-coupled Receptor 98
- Antigen symbol:GPR98
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Gorilla,Human
- Immunogen:Synthetic 20 amino acid peptide from human GPR98.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
GPR98 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 - 10 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GPR98
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human