Anti-SEMA4B Rabbit Polyclonal Antibody
Catalog # PRSI71-113
Supplier: ProSci Inc.
Specifications
- Antigen name:Semaphorin 4B
- Antigen symbol:SEMA4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Gorilla,Human
- Antigen synonyms:SemC|(Semaphorin) 4B|transmembrane domain (TM) and short cytoplasmic domain|immunoglobulin domain (Ig)|Sema domain|mKIAA1745|Semac
- Concentration:0.8 mg/mL
- Immunogen:Synthetic 19 amino acid peptide from internal region of human SEMA4B.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SEMA4B Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 15 - 20 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SEMA4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human