Anti-SLC1A5 Rabbit Polyclonal Antibody
Catalog # PRSI71-110
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 1 Member A5
- Antigen symbol:SLC1A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Gibbon,Human
- Concentration:0.3 mg/mL
- Immunogen:Synthetic 17 amino acid peptide from internal region of human SLC1A5.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC1A5 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 15 - 20 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC1A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human