Anti-CEBPB Mouse Monoclonal Antibody
Catalog # PRSI48-839
Supplier: ProSci Inc.
Specifications
- Antigen name:CCAAT/Enhancer-binding Protein Beta
- Antigen symbol:CEBPB
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TCF-5|Liver activator protein|LIP|ORF Names: PP9092|C/EBP beta|Transcription factor 5|LAP|TCF5|CEBPB|Liver-enriched inhibitory protein|Nuclear factor NF-IL6|CCAAT/enhancer-binding protein beta
- Immunogen:CEBPB monoclonal antibody was raised against amino acids 39 - 78 of CEBPB (Human).
- Size:50 µg
- Pk:50 µG
Specifications
About this item
NF-IL6 is a member of the CAAT/enhancer-binding protein (C/EBP) family of transcription factors. It has been shown to directly interact with the retinoblastoma (RB) protein.
Protein G Column
PBS, pH 7.4. No preservative added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CEBPB
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: