Anti-GPR132 Rabbit Polyclonal Antibody
Catalog # PRSI48-227
Supplier: ProSci Inc.
Specifications
- Antigen name:G Protein-coupled Receptor 132
- Antigen symbol:GPR132
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:G2A
- Immunogen:GPR132 antibody was raised against a peptide located in the C-Terminal domain of GPR132 (Human).
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
G2A belongs to the Lysophospholipid/Lysosphingolipid Receptor family and is an anti-proliferative cell cycle regulator that plays a critical role in controlling peripheral lymphocyte homeostasis. Loss of G2A expression leads to the development of a late-onset autoimmune syndrome (Le et al, 2001). Lysophosphatidylcholine (LPC) is the activating ligand for G2A.
GPR132 antibody can be used in ELISA, and immunohistochemistry starting at 28 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GPR132
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human