Order Entry
Italy
ContactUsLinkComponent
Anti-PRKAG3 Goat Polyclonal Antibody
Anti-PRKAG3 Goat Polyclonal Antibody
Catalog # PRSI46-941
Supplier:  ProSci Inc.
undefined
Anti-PRKAG3 Goat Polyclonal Antibody
Catalog # PRSI46-941
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 3
  • Antigen symbol:
    PRKAG3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Form:
    Liquid
  • Antigen synonyms:
    AMPKG3|AMPK gamma3|gamma-3 subunit|AMPK gamma-3 chain|5'-AMP-activated protein kinase
  • Concentration:
    500 ug/mL
  • Immunogen:
    PRKAG3 antibody was raised against a 14 amino acid synthetic peptide near the internal region of PRKAG3.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

PRKAG3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PRKAG3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PRKAG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0