Anti-APOL4 Goat Polyclonal Antibody
Catalog # PRSI46-807
Supplier: ProSci Inc.
Specifications
- Antigen name:Apolipoprotein L4
- Antigen symbol:APOL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:APOL-IV|APOLIV|FXI
- Concentration:500 ug/mL
- Immunogen:Apolipoprotein L4 antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of Apolipoprotein L4.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 37 kDa band observed in human placenta lysates (calculated MW of 38.7 kDa according to NP_085146.2). Recommended concentration: 0.3-1 μg/mL.
Apolipoprotein L4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Apolipoprotein L4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APOL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human