Order Entry
Italy
ContactUsLinkComponent
Anti-MUC13 Goat Polyclonal Antibody
Anti-MUC13 Goat Polyclonal Antibody
Catalog # PRSI45-915
Supplier:  ProSci Inc.
undefined
Anti-MUC13 Goat Polyclonal Antibody
Catalog # PRSI45-915
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Mucin 13 Cell Surface Associated
  • Antigen symbol:
    MUC13
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Form:
    Liquid
  • Antigen synonyms:
    cell surface associated|MUC-13|MUC13 Down regulated in colon cancer 1|MUC13_HUMAN.|RECC|DRCC1|UNQ6194|Mucin 13|MUC 13
  • Concentration:
    500 ug/mL
  • Immunogen:
    MUC13 antibody was raised against a 12 amino acid synthetic peptide near the internal region of MUC13.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

MUC13 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MUC13 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MUC13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: