Anti-AKR1B10 Goat Polyclonal Antibody
PRSI45-234
- Antigen name:Aldo-keto reductase family 1 member B10
- Antigen symbol:AKR1B10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:aldo-keto reductase family 1|ALDRLn|AKR1B11|ARL-1|small intestine reductase|AKR1B12|aldose reductase-like 1N: aldose reductase-like peptide|aldose reductase-related protein|AKR1B10|ARL1|member B10 (aldose reductase) |member B10|MGC14103|HIS|member B11 (aldose reductase-like)
- Concentration:500 ug/mL
- Immunogen:AKR1B10 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of AKR1B10.
- Size:100 µg
- Pk:100 µG
AKR1B10 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
AKR1B10 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: AKR1B10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human