Order Entry
Italy
ContactUsLinkComponent
Anti-MORF4L2 Goat Polyclonal Antibody
Anti-MORF4L2 Goat Polyclonal Antibody
  PRSI45-091
undefined
Anti-MORF4L2 Goat Polyclonal Antibody
  PRSI45-091

 

  • Antigen name:
    Mortality Factor 4 Like 1
  • Antigen symbol:
    MORF4L2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    MRGX|MORFL2
  • Concentration:
    500 ug/mL
  • Immunogen:
    MORF4L2 antibody was raised against a 14 amino acid synthetic peptide near the N-Terminus of MORF4L2.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

MORF4L2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MORF4L2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MORF4L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human