Anti-MAPK3 Goat Polyclonal Antibody
Catalog # PRSI45-087
Supplier: ProSci Inc.
Specifications
- Antigen name:Mitogen-Activated Protein Kinase 3
- Antigen symbol:MAPK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:HS44KDAP|ERK1|HUMKER1A|p44-MAPK|ERT2|PRKM3|P44ERK1|P44MAPK|ERK-1|p44-ERK1
- Concentration:500 ug/mL
- Immunogen:MAPK3 antibody was raised against a 12 amino acid synthetic peptide near the N-Terminus of MAPK3.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MAPK3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
MAPK3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human