Anti-LDLRAP1 Goat Polyclonal Antibody
Catalog # PRSI45-082
Supplier: ProSci Inc.
Specifications
- Antigen name:Low Density Lipoprotein Receptor Adaptor Protein 1
- Antigen symbol:LDLRAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ARH1|ARH2|FHCB1|FHCB2|ARH
- Concentration:500 ug/mL
- Immunogen:LDLRAP1 antibody was raised against a 13 amino acid synthetic peptide near the N-Terminus of LDLRAP1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
LDLRAP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
LDLRAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LDLRAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human