Order Entry
Italy
ContactUsLinkComponent
Anti-LCP2 Goat Polyclonal Antibody
Anti-LCP2 Goat Polyclonal Antibody
Catalog # PRSI45-081
Supplier:  ProSci Inc.
undefined
Anti-LCP2 Goat Polyclonal Antibody
Catalog # PRSI45-081
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Lymphocyte Cytosolic Protein 2
  • Antigen symbol:
    LCP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    76 kDa tyrosine phosphoprotein|lymphocyte cytosolic protein 2|SLP-76 tyrosine phosphoprotein|SH2 domain-containing leukocyte protein of 76 kDa|SH2 domain-containing leukocyte protein of 76kD
  • Concentration:
    500 ug/mL
  • Immunogen:
    LCP2 antibody was raised against an 11 amino acid synthetic peptide near the N-Terminus of LCP2.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

LCP2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

LCP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human