Anti-PRKAA2 Goat Polyclonal Antibody
Catalog # PRSI42-828
Supplier: ProSci Inc.
Specifications
- Antigen name:Protein Kinase AMP-activated Catalytic Subunit Alpha 2
- Antigen symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:AMPKa2|AMPK|AMPK2|PRKAA
- Concentration:500 ug/mL
- Immunogen:PRKAA2 antibody was raised against an 11 amino acid synthetic peptide near the internal region of PRKAA2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
PRKAA2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human