Anti-NAIP Goat Polyclonal Antibody
Catalog # PRSI42-537
Supplier: ProSci Inc.
Specifications
- Antigen name:NLR Family Apoptosis Inhibitory Protein
- Antigen symbol:NAIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Form:Liquid
- Antigen synonyms:psiNAIP|FLJ58811|FLJ42520|NLRB1|BIRC1|FLJ18088
- Concentration:500 ug/mL
- Immunogen:NAIP antibody was raised against a 14 amino acid synthetic peptide near the N-Terminus of NAIP.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
NAIP antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
NAIP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NAIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: