Anti-Tetanus Goat Polyclonal Antibody
PRSI35-381
- Antigen name:Tetanus
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Bacteria
- Western blot:Yes
- Form:Liquid
- Immunogen:Purified tetanus toxoid
- Size:1 mL
- Pk:1 mL
Purified IgG fraction of antiserum to a purity of >90%.
Tetanus toxoid antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Tetanus
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bacteria