Anti-Streptococcus Rabbit Polyclonal Antibody
Catalog # PRSI35-197
Supplier: ProSci Inc.
Specifications
- Antigen name:Streptococcus
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Bacteria
- Western blot:Yes
- Form:Liquid
- Immunogen:Streptococci, group A
- Size:1 mL
- Pk:1 mL
Specifications
About this item
Purified IgG fraction of antiserum to a purity of >90%.
Group A Streptococcus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Streptococcus
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria