Order Entry
Italy
ContactUsLinkComponent
Anti-TSPAN32 Rabbit Polyclonal Antibody
Anti-TSPAN32 Rabbit Polyclonal Antibody
Catalog # PRSI30-212
Supplier:  ProSci Inc.
undefined
Anti-TSPAN32 Rabbit Polyclonal Antibody
Catalog # PRSI30-212
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Tetraspanin 32
  • Antigen symbol:
    TSPAN32
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    PHMX|TSSC6|PHEMX|ART1
  • Immunogen:
    A synthetic peptide corresponding a region of human TSPAN32.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This gene is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. This gene may play a role in malignancies and disease that involve this region as well as hematopoietic cell function. This gene is described as a member of the tetraspanin superfamily.This gene is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. This gene is located among several imprinted genes; however, this gene, as well as the tumor-suppressing subchromosomal transferable fragment 4 (TSSC4), escapes imprinting. This gene may play a role in malignancies and disease that involve this region as well as hematopoietic cell function. This gene is described as a member of the tetraspanin superfamily. Alternative splicing of this gene results in several transcript variants; however, not all variants have been fully described.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TSPAN32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human