Anti-GPT Rabbit Polyclonal Antibody
PRSI30-103
- Antigen name:Glutamic-Pyruvate Transaminase
- Antigen symbol:GPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Glutamic--pyruvic transaminase 1|AAT1|GPT1|GPT|Alanine aminotransferase 1|Glutamic--alanine transaminase 1 |Glutamate pyruvate transaminase 1|ALT1|GPT 1
- Immunogen:A synthetic peptide corresponding a region of human GPT.
- Size:100 µg
- Pk:100 µG
GPT participates in cellular nitrogen metabolism and also in liver gluconeogenesis starting with precursors transported from skeletal muscles.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog