Order Entry
Italy
ContactUsLinkComponent
Anti-CDC23 Rabbit Polyclonal Antibody
Anti-CDC23 Rabbit Polyclonal Antibody
Catalog # PRSI29-826
Supplier:  ProSci Inc.
undefined
Anti-CDC23 Rabbit Polyclonal Antibody
Catalog # PRSI29-826
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Cell Division Cycle 23
  • Antigen symbol:
    CDC23
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    Cell Division Cycle protein 23 homolog|CUT23|APC8|Anaphase-promoting complex subunit 8|ANAPC8
  • Immunogen:
    A synthetic peptide corresponding a region of human CDC23.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

CDC23 shares strong similarity with Saccharomyces cerevisiae Cdc23, a protein essential for cell cycle progression through the G2/M transition. This protein is a component of anaphase-promoting complex (APC), which is composed of eight protein subunits and highly conserved in eukaryotic cells. APC catalyzes the formation of cyclin B-ubiquitin conjugate that is responsible for the ubiquitin-mediated proteolysis of B-type cyclins. This protein and 3 other members of the APC complex contain the TPR (tetratricopeptide repeat), a protein domain important for protein-protein interaction.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDC23
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: