Order Entry
Italy
ContactUsLinkComponent
Anti-SLC22A1 Rabbit Polyclonal Antibody
Anti-SLC22A1 Rabbit Polyclonal Antibody
Catalog # PRSI29-592
Supplier:  ProSci Inc.
undefined
Anti-SLC22A1 Rabbit Polyclonal Antibody
Catalog # PRSI29-592
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Solute Carrier Family 22 Member A1
  • Antigen symbol:
    SLC22A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    HOCT1|SLC22A1|OCT1|oct1_cds
  • Immunogen:
    A synthetic peptide corresponding a region of human SLC22A1.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. SLC22A1 contains twelve putative transmembrane domains and is a plasma integral membrane protein.Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC22A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat