Specifications
- Antigen name:Pre-mRNA Processing Factor 6
- Antigen symbol:PRPF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:S. cerevisiae|androgen receptor N-terminal domain transactivating|hPrp6|U5-102K|PRP6 pre-mRNA processing factor 6 homolog|C20orf14|TOM|ANT-1|U5 snRNP-associated 102 kDa protein
- Immunogen:A synthetic peptide corresponding a region of human PRPF6.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
PRPF6 appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. PRPF6 also can bind androgen receptor, providing a link between transcriptional activation and splicing.The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PRPF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat