Specifications
- Antigen name:Zinc Finger Protein 76
- Antigen symbol:ZNF76
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ZNF76.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ZNF76 belongs to the krueppel C2H2-type zinc-finger protein family. ZNF76 is a general transcription repressor targeting TATA-binding protein (TBP), through a process regulated by sumoylation. ZNF76 interacts with TBP through both its N and C termini, and both regions are required for ZNF76 to exert its inhibitory function on p53-mediated transactivation. As a TBP-interacting transcriptional modulator, ZNF76 is also regulated by alternative splicing.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZNF76
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat