Anti-FOXK1 Rabbit Polyclonal Antibody
Catalog # PRSI28-707
Supplier: ProSci Inc.
Specifications
- Antigen symbol:FOXK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Myocyte nuclear factor|Forkhead box protein K1|MNF|Fork head|FOXK1|FOXJ2 forkhead factor|FHX
- Immunogen:A synthetic peptide corresponding a region of mouse FOXK1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Foxk1 is a transcriptional regulator that binds to the upstream enhancer region (CCAC box) of myoglobin gene. It has a role in myogenic differentiation and in remodeling processes of adult muscles that occur in response to physiological stimuli.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FOXK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse